Learn More
Buerkle™ SteriPlast™ Sampling Spoons
Sample powders, granules, and pastes. Buerkle™ SteriPlast™ Sampling Spoons are made of polystyrene and come sterile and individually wrapped.
697.00 NOK - 738.00 NOK
Specifications
Product Type | Spoon |
---|---|
Material | Polystyrene |
Autoclavable | Not Autoclavable |
Sterility | Sterile |
Color | White |
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
15356327
![]() ![]() |
Buerkle™
5378-1011 |
2.5 mL | 127 mm |
697.00 NOK
Pack of 100 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||||
15351766
![]() ![]() |
Buerkle™
5378-1012 |
10 mL | 170 mm |
738.00 NOK
Pack of 100 |
Estimated Shipment: 20-06-2025
Log in to see stock. |
|||||
Description
- Intended for sampling of powders, granules, and pastes
- Sterilized by gamma radiation
- Individually packaged for one-way needs
- Production, assembling, and packaging according to cleanroom class 7 (10.000)
Specifications
Spoon | |
Not Autoclavable | |
White |
Polystyrene | |
Sterile | |
Sampling Powders, Granules, and Pastes |
Product Title
Buerkle™ SteriPlast™ Sampling Spoons