Learn More
Fisherbrand™ Sterile Syringes for Single Use
Manual syringe without needle for research purposes only. Suitable for use with syringe filters
Brand:  Fisherbrand™ 14955450
 View more versions of this product
Product Code. 15889142
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • GAMMA sterilization
  • Sterile
  • Individually wrapped in a paper/plastic blister pack
Services
5 Years
Specifications
Specifications
3 parts
1 mL
Sterile
Slip Tip
Leur Slip
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Sterile Syringes for Single Use > 1mL