Learn More
Fisherbrand™ Sterile Syringes for Single Use
Manual syringe without needle for research purposes only. Suitable for use with syringe filters
Brand: Fisherbrand™ 14955459
Description
- GAMMA sterilization
- Sterile
- Individually wrapped in a paper/plastic blister pack
Services
5 Years
Specifications
14.85 mm | |
10 mL | |
Sterile |
Luer-Lock | |
Luer Lock |
Plastic syringes
File size: 174.8 kB
Product Content Correction
Product Title
Fisherbrand™ Sterile Syringes for Single Use > 10mL