Learn More
Bochem™ Steel Wire Mesh
Brand:  Bochem™ 12751
 View more versions of this product
Product Code. 10410353
  • 34.60 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Specifications
85 mm
120 x 120 mm
Square
Wire Gauze with Ceramic Center
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Steel Wire Mesh > Dimensions (L x W): 120 x 120mm