Learn More
Fisherbrand™ Steel Stand Rings with Rod
Provides for ease of connection on ring stands. Fisherbrand™ Steel Stand Rings with Rod are manufactured from plated mild steel.
Brand:  Fisherbrand™ 2010/17
 View more versions of this product
Product Code. 12986836
  • 148.00 NOK / Each
Only null left
Estimated Shipment: 22-07-2025
Add to basket
Description
Description
  • 8mm diameter stems
  • 140mm stem lengths
  • Rings in a variety of diameters
Specifications
Specifications
Steel
150 mm
Stand Ring
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Steel Stand Rings with Rod > Inner Diameter: 150mm