Learn More
Fisherbrand™ Steel Stand Rings with Rod
Provides for ease of connection on ring stands. Fisherbrand™ Steel Stand Rings with Rod are manufactured from plated mild steel.
Brand:  Fisherbrand™ 2010/14
 View more versions of this product
Product Code. 12976836
  • 152.00 NOK / Each
Only null left
Estimated Shipment: 12-08-2025
Add to basket
Description
Description
  • 8mm diameter stems
  • 140mm stem lengths
  • Rings in a variety of diameters
Specifications
Specifications
Steel
127 mm
Stand Ring
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Steel Stand Rings with Rod > Inner Diameter: 127mm