Learn More
asecos™ Steel Grid Storage Level
The Steel Grid Storage Level provides an additional storage level.
1025.00 NOK - 1470.00 NOK
Product Code | Brand | Weight (Metric) | Depth (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Depth (Metric) | Price | Quantity & Availability | ||||
17836376
|
asecos™
27877 |
8 kg | 400.00 mm |
1040.00 NOK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
17876366
|
asecos™
27885 |
8 kg | 600.00 mm |
1470.00 NOK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
17836366
|
asecos™
27878 |
11 kg | 600.00 mm |
1265.00 NOK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
17886396
|
asecos™
27880 |
12 kg | 600.00 mm |
1120.00 NOK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
17846536
|
asecos™
27879 |
14 kg | 400.00 mm |
1025.00 NOK
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
Description
- Incl. reinforcing traverses and locking pins
- Galvanised steel
Specifications
Steel | |
Storage Level |