Learn More
R&L Enterprises™ Gas Deflagrating Spoons
Excellent for deflagration demonstration purposes. R&L Enterprises™ Gas Deflagrating Spoons are comprised of steel with an aluminum flange.
Brand:  R&L Enterprises™ Z4604
Product Code. 12965941
  • 444.00 NOK / Pack of 6
Only null left
Estimated Shipment: 04-09-2025
Add to basket
Description
Description
  • Durable construction
  • Supplied in packs of 6
Specifications
Specifications
Deflagrating Spoon
Steel
6 Pk.
400 mm
Cork and Cap
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&L Enterprises™ Gas Deflagrating Spoons > Length: 400mm