Learn More
Justrite™ Steel Bench Can
937.38 NOK - 1205.00 NOK
Specifications
Material Steel
Capacity (Metric) 4 L
View More Specs
Products 2
Product Code Brand Height (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Price Quantity & Availability  
11953629
View Documents
Justrite™
10291
267 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11963629
View Documents
Justrite™
10385
114 mm
1205.00 NOK
Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
Steel
4 L
Documents
Product Certifications
Product Content Correction
Product Title
Justrite™ Steel Bench Can