Learn More
Brady™ Static Dissipative Thermal Transfer Printable Labels
Printable, static dissipative polyester labels for Thermal Transfer Printers.
Brand: Brady™ 029913
This item is not returnable.
View return policy
Description
- Material is UL and CSA approved with its respective ribbon
- Ideal for Circuit Board Labeling
- For smooth surfaces
Specifications
Not Autoclavable | |
White | |
Die-cut | |
Thermal Transfer | |
Non-sterile | |
BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500 | |
3 | |
THT-2-473-10 | |
10,000 Labels | |
22.86 mm (W) x 6.35 mm (H) | |
-40°C to +120°C | |
0.09 mm | |
22.86 mm (W) x 6.35 mm (H) | |
22.9 mm | |
22.9 mm | |
6.4 mm | |
THT-2-473-10 |
Permanent Acrylic | |
Gloss | |
Static-dissipative Polyester | |
Labels | |
Circuit Board Labeling, Electrical Labeling, General Identification | |
28.6 mm | |
-40°C | |
10000 Label(s) / Roll | |
Rectangle | |
-40°F to 356°F | |
0.0034 in | |
8.9 mm | |
0.9 in | |
22.86 mm | |
0.25 in | |
6.4 mm |
Product Content Correction
Product Title
Brady™ Static Dissipative Thermal Transfer Printable Labels > Static Dissipative Thermal Transfer Printable Labels; Labels per Row: 3; Shape: Rectangle; Height (Metric): 6.4 mm; Vertical Repeat (Metric): 8.9 mm; Width (Metric): 22.9 mm