Learn More
Spectrum™ Labs Standard RC Dialysis Disc
For standard grade RC dialysis tubing
Brand:  Spectrum™ Labs 132482
2936.39 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12727526
  • 3015.00 NOK / Pack of 50
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
  • Regenerated cellulose (RC) precut membrane discs
  • Supplied dry with a glycerol as a humectant
Specifications
Specifications
Standard RC Dialysis Disc
100 mm
50 Pk.
For standard grade RC dialysis tubing
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Spectrum™ Labs Standard RC Dialysis Disc > Diameter: 100mm