Learn More
Koettermann™ Standard Hanging Cabinet, 900x480x366mm
Koettermann™ Standard Hanging Cabinet, 900x480x366mm provides suitable storage to accommodate a variety of laboratory needs.
5860.00 NOK - 6210.00 NOK
Specifications
Dimensions (D x W x H) | 366 x 900 x 480 mm |
---|---|
No. of Doors | 2 |
No. of Shelves | 1 |
Product Type | Wall-mounted Cabinet |
Product Code | Brand | Locking Mechanism | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Locking Mechanism | Price | Quantity & Availability | |||||
15743112
|
Koettermann™
307-00198 |
Yes |
6210.00 NOK
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
15733112
|
Koettermann™
307-00197 |
No |
5860.00 NOK
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
Descripción
- Wall-mounted cabinet
- Two doors
- One shelf
- Side panels
Especificaciones
366 x 900 x 480 mm | |
2 | |
1 | |
Wall-mounted Cabinet |