Learn More
Buerkle™ Worm-Threaded Hose Clips
Manufactured from stainless steel 1.4016.
Brand:  Buerkle™ 8677-1625
219.40 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10550784
  • 261.00 NOK / Pack of 10
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Description
  • With screw thread
  • Screw is zinc-coated
Specifications
Specifications
16 to 27 mm
Hoses
16 to 27 mm dia.
Stainless Steel
10/Pk.
Worm-threaded Hose Clip
Documents
One moment while we fetch your results.
Product Title
Buerkle™ Worm-Threaded Hose Clips > Diameter: 16 to 27mm