Learn More
Sartorius™ Stainless Steel Weighing Scoop
Weighing scoop for use with laboratory balances.
Brand: Sartorius™ 641214
Description
- Weighing scoop of chrome nickel steel
Specifications
Weighing Scoop | |
Autoclavable | |
90 x 32 x 8 mm |
Stainless Steel | |
Non-sterile |