Learn More
S Murray and Co™ Stainless Steel Watchmaker Forceps
Use these forceps for dissection and manipulating slides and cover glasses. S Murray™ Stainless Steel Watchmaker Forceps are made from high grade stainless steel in accordance to the specifications of ISO 7153-1.
96.25 NOK - 101.90 NOK
Specifications
Product Type | Forceps, Watchmaker |
---|---|
Material | Stainless Steel |
For Use With (Application) | For dissection |
Quantity | Each |
Length (Metric) | 110 mm |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12311379
|
S Murray and Co™
E010/04 |
No. 4 |
101.90 NOK
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
12740926
|
S Murray and Co™
E010/07 |
No. 7 |
96.25 NOK
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
Description
- Stainless Steel forceps
- For dissection
Specifications
Forceps, Watchmaker | |
For dissection | |
110 mm |
Stainless Steel | |
Each |
Product Title
S Murray and Co™ Stainless Steel Watchmaker Forceps