Learn More
VACUUBRAND™ Stainless Steel Vacuum Tubing With Small Flanges KF
Providing durable connection solutions for the laboratory.
1215.00 NOK - 1715.00 NOK
Specifications
Product Type | Vacuum Tubing |
---|---|
Material | Stainless Steel |
Packaging Quantity | 1 Each |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
10360692
|
VACUUBRAND™
20673000 |
60mm |
1215.00 NOK
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
10390352
|
VACUUBRAND™
20673015 |
100mm |
1505.00 NOK
Each |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
10476641
|
VACUUBRAND™
20673016 |
130mm |
1715.00 NOK
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
10670862
|
VACUUBRAND™
20673014 |
80mm |
1235.00 NOK
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
Description
- Manufactured from stainless steel
- Flange and tube TIG welded
- Available in a range of sizes
Specifications
Vacuum Tubing | |
Stainless Steel | |
1 Each |