Learn More
Thermo Scientific™ Stainless Steel Unions, Tees and Crosses
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Brand: Thermo Scientific™ U-412
688.82 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
721.61 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
- Absolute zero or low dead volume formats
- Includes two stainless steel nuts and ferrules
Specifications
Union | |
Union with 2 stainless steel nuts and ferrules | |
Stainless Steel |
0.129 ÎĽL | |
0.020 in. through hole | |
Waters Fittings |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13