Learn More
Sampling Systems™ Stainless Steel Tweezers
Essential equipment for laboratories. Sampling Systems™ Stainless Steel Tweezers are made from hygienic 304 stainless steel.
Brand:  Sampling Systems™ A394-300
Product Code. 15361851
  • 115.44 NOK / Each
Only null left
Estimated Shipment: 18-07-2025
Description
  • Polished finish
  • Range of sizes
  • 304 stainless steel
Specifications
Specifications
Stainless Steel
300 mm
Tweezers
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ Stainless Steel Tweezers > Length: 300mm