Learn More
Bochem™ Stainless Steel Tubing Clips
Used to control the flow of liquids. Bochem™ Stainless Steel Tubing Clips are manufactured from stainless steel.
Brand:  Bochem™ 12135
 View more versions of this product
Product Code. 10306101
  • 45.20 NOK / Each
Only null left
Estimated Shipment: 22-10-2025
Description
  • Manufactured from stainless steel
  • Used with tubing
Specifications
Specifications
25 mm
Stainless Steel
Tubing Clip
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Stainless Steel Tubing Clips > Diameter: 25mm