Learn More
Bochem™ Stainless Steel Tubing Clip
Manufactured from 18/10 stainless steel. Bochem™ Stainless Steel Tubing Clip are used with tubing.
Brand: Bochem™ 12133
This item is not returnable.
View return policy
Description
- Made from 18/10 stainless steel
- For use with tubing
Specifications
17 mm | |
Stainless Steel (18/10) | |
1 Pack | |
Tubing Clip |
Product Content Correction
Product Title
Bochem™ Stainless Steel Tubing Clip > Diameter: 17mm