Learn More
Bochem™ Stainless Steel Tube Clamp
Manufactured from stainless steel. BOCHEM™ Stainless Steel Tube Clamps are self-tensing.
Brand:  Bochem™ 2291
Product Code. 10453151
  • 131.00 NOK / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Description
  • Self-tension
  • Ridges at the top
Specifications
Specifications
140 mm
Stainless Steel
Tube Clamp
Documents
One moment while we fetch your results.
Product Title
Bochem™ Stainless Steel Tube Clamp > Length: 140mm