Learn More
Bochem™ Stainless Steel Transport Container with Lid
Click to view available options
Capacity (Metric):
75 L
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 400mm |
| Height (Metric) | 375H |
| Capacity (Metric) | 75 L |
| Material | Stainless Steel (18/10) |
| Product Type | Container/Lids |
Product Content Correction
Product Title
Bochem™ Stainless Steel Transport Container with Lid >