Learn More
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ASTM E 11
Brand:  RETSCH 60.117.000500
 View more versions of this product
Product Code. 10600452
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
1 Each
Test Sieve
Shaker AS 200
40mm
150 mm
Stainless Steel
500 ÎĽm
ASTM E 11
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ASTM E 11 > Pore Size: 500 ÎĽm