Learn More
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ASTM E 11
Brand:  RETSCH 60.117.000425
 View more versions of this product
Product Code. 10064761
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
1 Each
Test Sieve
Shaker AS 200
40mm
150 mm
Stainless Steel
425 ÎĽm
ASTM E 11
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ASTM E 11 > Pore Size: 425 ÎĽm