Learn More
Endecotts™ Stainless Steel Test Sieve
Woven wire test sieves for all types of laboratory sampling and particle size analysis
Brand: Endecotts™ 008SAW.063H
This item is not returnable.
View return policy
Description
- Precision frame (ensures consistent nestability
- Precise aperture (in accordance with ISO 3310-1, ASTM E11 or other specifications)
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)
Specifications
63 μm | |
8 in. | |
Woven Stainless Steel Wire |
ASTM E11 | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Endecotts™ Stainless Steel Test Sieve > Pore Size: 63 μm