Learn More
Endecotts™ Stainless Steel Test Sieve
Woven wire test sieves for all types of laboratory sampling and particle size analysis
Brand:  Endecotts™ 008SAW.063H
 View more versions of this product
Product Code. 11542065
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Precision frame (ensures consistent nestability
  • Precise aperture (in accordance with ISO 3310-1, ASTM E11 or other specifications)
  • Natural fillet (free flowing of sample)
  • Totally sealed (no crevice to lose material)
  • Evenly tensioned mesh (ensures accurate analysis)
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number (ensures full traceability)
Specifications
Specifications
63 μm
8 in.
Woven Stainless Steel Wire
ASTM E11
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Endecotts™ Stainless Steel Test Sieve > Pore Size: 63 μm