Learn More
Nickel Electro™ Stainless Steel Gable Lid
Maintain stability in bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Gable Lid minimises heat loss and reduces evaporation.
Brand:  Nickel Electro™ SL4-22
 View more versions of this product
Product Code. 11479519
  • 2590.00 NOK / Each
Only null left
Estimated Shipment: 21-08-2025
Description
  • Stainless steel
  • Insulated handle
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
Specifications
Stainless steel gable lid
22 L Stirred Bath
Stainless Steel
3 Years
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Gable Lid > For Use With: 22L Bath