Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN30, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ UN30B02070
Product Code. 17570076
  • 913.00 NOK / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
UN30 Oven
Drip Tray
Stainless Steel
Rim: 15 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN30, 15 mm > For Use With: UN30