Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF750, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand: Memmert™ SF750B02075
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
SF750 Sterilizer | |
Drip Tray | |
Rim: 15 mm | |
Stainless Steel |