Learn More
Bochem™ Stainless Steel Sharp Tip Forceps
Constructed from stainless steel. Bochem™ Stainless Steel Sharp Tip Forceps have a variety of lab applications.
Brand:  Bochem™ 1132
 View more versions of this product
Product Code. 10274031
  • 71.90 NOK / Each
Only null left
Estimated Shipment: 16-07-2025
Description
  • With guide-pin
  • Sharp tip
  • Stainless steel
Specifications
Specifications
Sharp
Stainless Steel
Forceps
Forceps
1 Pack
130 mm
Guide pin
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Stainless Steel Sharp Tip Forceps > Length: 130mm