Learn More
RETSCH Stainless-Steel Seven Analysis Test Sieves, 200 mm Dia.
Shop All Retsch Products
Click to view available options
Pore Size:
63 μm
90 μm
1 mm
125 μm
180 μm
250 μm
355 μm
500 μm
630 μm
710 μm
Unit Size:
Each
Tekniske data
Tekniske data
Quantity | 1 Each |
Pore Size | 180 μm |
Type | Test Sieve |
Certifications/Compliance | ISO 3310 and ASTM E 11 |
Height (Metric) | 25mm |
For Use With (Application) | Separation, fractioning, particle size determination |
Diameter (Metric) Outer | 200 mm |
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
RETSCH Stainless-Steel Seven Analysis Test Sieves, 200 mm Dia. >
Ser du en mulighed for forbedring?Del en indholdskorrektion