Learn More
Fisherbrand™ Stainless Steel Round Immersion Basket with Handles
Facilitates complete immersion within S 50 R Ultrasonic Cleaner baths. Fisherbrand™ Stainless Steel Round Immersion Baskets with Handles feature a functional and modern design.
Brand: Fisherbrand™
Additional Details : Weight : 0.50000kg
Description
- Mesh bottom
- Two handles, one on each side
- Handles are coated with plastic
Specifications
Basket | |
220 mm | |
70 mm |
575 mm | |
Ultrasonic Unit, Elmasonic S 50 R |