Learn More
Julabo™ Stainless Steel/PTFE External Pt100 Sensor
Brand:  Julabo™ 8981014
 View more versions of this product
Product Code. 11994768
  • 10500.00 NOK / Each
Only null left
Estimated Shipment: 04-07-2025
Specifications
External Pt100 Sensor
PTFE-coated Stainless Steel
1200 x 6 mm
Suitable for CF31, CF41, F25-HE, F25-HL, F25-ME, F26-ME Refrigerated/Heating Circulator
1200 x 6 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Julabo™ Stainless Steel/PTFE External Pt100 Sensor > Size: 1200 x 6mm