Learn More
Nickel Electro™ Stainless Steel Perforated Tray
Nickel Electro™ Stainless Steel Perforated Tray is to be used with flask clips and can be used in a variety of baths.
Brand: Nickel Electro™ SFT0025
This item is not returnable.
View return policy
Description
- Supplied ready to assemble
- Stainless steel construction
- Suits glass/polypropylene Erlenmeyer flasks
- Tray simply drops onto shaker or rocking table platform and locates snugly
Specifications
25 mL flask | |
Stainless Steel | |
NE5-28 Shaker Baths |
Perforated Tray | |
Yes | |
39 |
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Perforated Tray > Capacity: 39 x 25mL