Learn More
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Brand:  Sartorius™ YCW522-00
3252.26 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10032071
  • 3340.00 NOK / Each
Estimated Shipment: 17-06-2025
Description
Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wood
200 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate > Mass: 200g