Learn More
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW512-02
3696.81 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10377441
  • 3795.00 NOK / Each
Only null left
Estimated Shipment: 19-06-2025
Specifications
Wood
100 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight With DAkkS Calibration Certificate > Mass: 100g