Learn More
Sartorius™ Stainless Steel OIML Class E2 Calibration Weight Without DAkkS Calibration Certificate
Highly polished steel calibration weights from 1g to 2kg
Brand:  Sartorius™ YCW6128-00
5346.34 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10238180
  • 5485.00 NOK / Each
Only null left
Estimated Shipment: 22-07-2025
Description
To OIML Class E2, F1 and F2 specification.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plastic
1 kg
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E2 Calibration Weight Without DAkkS Calibration Certificate > Mass: 1kg