Learn More
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW511-02
8481.62 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10797612
  • 8700.00 NOK / Each
Estimated Shipment: 26-06-2025
Explore more special offers
Specifications
Wood
100 g
Product Title
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight With DAkkS Calibration Certificate > Mass: 100g