Learn More
Bochem™ Stainless Steel Microscopy Scissors
Manufactured from stainless steel. Bochem™ Stainless Steel Microscopy Scissors come in three handle types.
Brand:  Bochem™ 4070
 View more versions of this product
Product Code. 10065090
  • 64.50 NOK / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Description
  • Manufactured from stainless steel
  • Three types: round-round, sharp-round, sharp-sharp
Specifications
Specifications
Microscopy Scissors
100 mm
Sharp/Sharp
Straight
Stainless Steel
Product Title
Bochem™ Stainless Steel Microscopy Scissors > Length/Tip Style: 100mm/Sharp/Sharp