Learn More
Bochem™ Stainless Steel Lid
Brand:  Bochem™ 8334
 View more versions of this product
Product Code. 10107440
  • 621.00 NOK / Each
Only null left
Estimated Shipment: 26-09-2025
Specifications
15 L Graduated Bucket
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Stainless Steel Lid > For use with: For 15L graduated bucket