Learn More
Bochem™ Stainless Steel Laboratory Scissors
Manufactured from stainless steel
Brand: Bochem™ 4001
This item is not returnable.
View return policy
Description
- Three types: round-round, sharp-round, and sharp-sharp
- Stainless steel
Specifications
Laboratory Scissors | |
115 mm | |
Stainless Steel | |
Sharp/Round |
Product Content Correction
Product Title
Bochem™ Stainless Steel Laboratory Scissors > Length/Tip Style: 115mm/Sharp/Round