Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
Brand:  Nickel Electro™ 204
 View more versions of this product
Product Code. 11419569
  • 728.00 NOK / Each
Only null left
Estimated Shipment: 21-08-2025
Description
  • Durable stainless steel laboratory beaker
  • 304 spec stainless steel beakers 
  • Will not shatter or chip
  • Suitable for laboratory applications since 
  • All surfaces are non-reacting 
  • Offer zero absorption
Specifications
Specifications
75 mm
Stainless Steel
Spout
700 mL
172 mm
Beaker
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1429373 Certificate TypeCertificate of Compliance Date21/05/2025 Catalog Number204
Lot Number1424043 Certificate TypeCertificate of Compliance Date30/03/2025 Catalog Number204
Lot Number1423664 Certificate TypeCertificate of Compliance Date26/03/2025 Catalog Number204
Lot Number1423662 Certificate TypeCertificate of Compliance Date23/02/2025 Catalog Number204
Lot Number1423663 Certificate TypeCertificate of Compliance Date21/02/2025 Catalog Number204
Promotions
Promotions
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Beakers with Spout > Capacity: 0.7L