Learn More
Welch Ilmvac™ Stainless Steel KF Reducing Adapter
Designed for use with Welch Ilmvac™ Rotary Vane Pumps.
Brand:  Welch Ilmvac™ 701401
 View more versions of this product
Product Code. 10657852
  • 412.00 NOK / Each
Only null left
Estimated Shipment: 10-07-2025
Description
  • Stainless Steel KF Reducing Adapter, DN16/25
  • Connect KF 25 for intake or exhaust with CRVpro
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
1 Each
DN 25/16
KF Reducing Adapter
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch Ilmvac™ Stainless Steel KF Reducing Adapter > Size: DN 25/16