Learn More
Welch Ilmvac™ Stainless Steel KF Reducing Adapter
Designed for use with Welch Ilmvac™ Rotary Vane Pumps.
Brand:  Welch Ilmvac™ 701405
 View more versions of this product
Product Code. 10591593
  • 554.00 NOK / Each
Only null left
Estimated Shipment: 10-07-2025
Description
  • Stainless Steel KF Reducing Adapter
  • Available in a variety of sizes
  • Connect cold trap for intake or exhaust Welch vacuum pumps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
1 Each
DN 40/25
KF Reducing Adapter
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch Ilmvac™ Stainless Steel KF Reducing Adapter > Size: DN 40/25