Learn More
Memmert™ Stainless Steel Grid, Electropolished
Additional electropolished stainless steel grid
Brand: Memmert™ HPP260ecoE28891
1558.43 NOK valid until 2025-04-30
Use promo code "24235" to get your promotional price.
This item is not returnable.
View return policy
Description
- Material: Stainless Steel
- Electropolished
Specifications
HPP260eco | |
Stainless Steel |
Special Offers
-
Multiple Manufacturers
Equipment Monthly Promotion
Discover exclusive monthly offers on scientific equipment to enhance your laboratory's capabilities.Promo Code: 24235Expires: 2025-04-30
New Offer Save Up to 50%
Product Content Correction
Product Title
Memmert™ Stainless Steel Grid, Electropolished > HPP260eco