Learn More
OHAUS™ Stainless Steel Flask Clamp
OHAUS™ Stainless Steel Flask Clamp is designed to securely grip and position various sizes of flasks.
Brand: OHAUS™ 30400086
349.55 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of stainless steel
- For use with flasks
- Designed to securely hold flask in place
Specifications
Flask Clamp | |
50 mL Flask | |
Silver |
Stainless Steel | |
Flask Clamp, 50 mL | |
Four Interlocking |
Product Content Correction
Product Title
OHAUS™ Stainless Steel Flask Clamp > For Use With: 50 mL Flask