Learn More
Cole-Parmer™ Stainless Steel Electronic Balance Weight
Brand:  Cole-Parmer™ 11956-20
Product Code. 15362294
  • 1895.00 NOK / Each
Only null left
Estimated Shipment: 10-12-2025
Specifications
Electronic Balance Weight
Stainless Steel
100 g
1 Pack
NIST/NVLAP
Polycarbonate
For Calibration of Electronic Balances and Routine Weighing Applications
1 Pack
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Stainless Steel Electronic Balance Weight > Mass: 100g