Learn More
Bochem™ Stainless Steel Container
Bochem™ Stainless Steel Container is constructed of Stainless Steel and is equipped with handles on the side.
Brand:  Bochem™ 8346
 View more versions of this product
Product Code. 10020811
  • 13540.00 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Description
  • Stainless Steel
  • Includes handles
  • Compatible lid sold separately
Specifications
Specifications
400mm
600mm
75 L
Stainless Steel (18/10)
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Stainless Steel Container > Capacity: 75L