Learn More
Lenz Laborglasinstrumente™ Stainless Steel Clips
52.60 NOK - 638.00 NOK
Specifications
Quantity | 10/Pk. |
---|---|
Type | Joint Clip |
Material | Stainless Steel |
For Use With (Equipment) | For conical ground joints |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11746974
|
Lenz Laborglasinstrumente™
01103214 |
Conical Ground Joint: NS 14/23 |
440.96 NOK
Pack of 10 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
10708841
|
Lenz Laborglasinstrumente™
01103229 |
Conical Ground Joint: NS 29/32 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12333840
|
Lenz Laborglasinstrumente™
01103229 |
Conical Ground Joint: NS 29/32 |
638.00 NOK
Pack of 10 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
Specifications
10/Pk. | |
Joint Clip | |
Stainless Steel | |
For conical ground joints |