Learn More
3M™ Stainless Steel Cleaner and Polish Aerosol
3M™ Stainless Steel Cleaner and Polish Aerosol protects all stainless steel surfaces in one step.
Brand:  3M™ YP208061744
Product Code. 15836234
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Polishes and protects all stainless steel surfaces in one step
  • The spray clings to vertical surfaces and leaves a light, non-greasy film that resist fingerprints and marking
  • Does not contain harmful chemicals and safe to use in food preparation areas
  • Water based with a pleasant citrus fragrance
Certificates
Promotions
Promotions
Product Content Correction
Product Title
3M™ Stainless Steel Cleaner and Polish Aerosol > Volume: 600 mL