Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F1, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F1 Tolerances
540.00 NOK - 19075.00 NOK
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
16692422
|
Ohaus™
80780331 |
1 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16602432
|
Ohaus™
80780332 |
2 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16612432
|
Ohaus™
80780333 |
5 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16622432
|
Ohaus™
80780334 |
10 mg |
540.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16632432
|
Ohaus™
80780335 |
20 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16642432
|
Ohaus™
80780336 |
50 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16652432
|
Ohaus™
80780337 |
100 mg |
540.00 NOK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
16662432
|
Ohaus™
80780338 |
200 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16672432
|
Ohaus™
80780339 |
500 mg |
540.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16682432
|
Ohaus™
80780340 |
1 g |
889.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16692432
|
Ohaus™
80780341 |
2 g |
877.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16602442
|
Ohaus™
80780342 |
5 g |
909.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16612442
|
Ohaus™
80780343 |
10 g |
957.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16622442
|
Ohaus™
80780344 |
20 g |
1025.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16632442
|
Ohaus™
80780345 |
50 g |
1110.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16642442
|
Ohaus™
80780346 |
100 g |
1345.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16652442
|
Ohaus™
80780347 |
200 g |
1600.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16662442
|
Ohaus™
80780348 |
500 g |
2280.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16672442
|
Ohaus™
80780349 |
1 kg |
3035.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16682442
|
Ohaus™
80780350 |
2 kg |
4670.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16692442
|
Ohaus™
80780351 |
5 kg |
6845.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
16602452
|
Ohaus™
80780352 |
10 kg |
12180.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
16612452
|
Ohaus™
80780353 |
20 kg |
19075.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
Description
- Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
Protective Case |
Product Title
Ohaus™ Stainless Steel Calibration Weights - OIML F1, NVLAP